Clamping Systems and Support Stands, Arrow
Supplier: Arrow Engineering
PHC-625 PHC-750 SC-750 ASE-625 ASE-750 SSS-15 SSS-55
59025-160EA 241.8 CAD
59025-160 59025-162 59025-164 59025-166 59025-168 59025-170 59025-172
Clamping Systems and Support Stands, Arrow
Clamps General Purpose Clamps
“Power Hold” Universal Clamps can be used for most applications. Designed with large knobs for powerful gripping, these clamps feature an adjustable stop for speed and safety in set up and operation. Made with precision machined aluminum rod for strength and durability. Can hold any stirrer that has a mounting rod stirring stand diameter from 9.5 – 16 mm (3/85/8") or from 12 – 19 mm (1/23/4").
Universal Swivel Clamp allows stirrer to be set at any compound angle for best stirring action
Arrow Support Stand consists of heavy duty steel base and stainless steel support rod. Base hasMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable