Dry Blending Container, Waring
Supplier: Waring Products
SS115
58983-100EA 2380.78 CAD
58983-100
Dry Blending Container, Waring
Homogenizers Standard Laboratory Blenders
Unique blade configuration designed for pulverizing and homogenizing dry substances
Note: Blending such as soil processing requires the use of a larger motor unit.
One liter stainless steel container and lid. Fits 1L Waring® blenders.
Order Now
SPECIFICATIONS
Environmentally Preferable