Vortex Genie® Mixers, Scientific Industries
Supplier: Scientific Industries
SI-0510 503-0277-00 SI-0511 SI-0570 SI-0136 SI-T236 580-2013-00 504-0235-00 504-0234-00 146-3011-00 146-6005-00 OK-0500-901 504-0039-00 503-0282-00 504-0233-00 SI-0236 SI-A236 SI-H506 SI-H512 SI-H524 SI-V506 SI-V512 SI-V524 SI-V525 503-0280-00 503-0279-00 503-0278-00 568-0001-00 503-0283-00 SI-0564 0K-0236-408 0K-0500-902 SI-V203 SIV206 SIV209 504-0232-00 0K-0500-901 0K-0513-900 SI-V203 SI-V206 SI-V209 SI-0513
13500-962EA 158.41 CAD
13500-962 13502-386 13502-388 13502-390 14216-184 14216-188 58815-180 58815-185 58815-206 58815-210 58815-212 58815-216 58815-222 58815-224 58815-230 58815-234 89037-464 97060-174 97060-176 97060-178 97060-180 97060-182 97060-184 97060-186 58815-184 58815-186 58815-188 58815-218 58815-220 89202-322 89202-338 14221-972 89401-416 89401-418 89401-420 89415-150 490010-010 490010-016 490010-018 490010-026 490010-036 490010-038 490010-040 490014-806
Vortex Genie® Mixers, Scientific Industries
Shakers Vortex Mixers
Vortex Genie® Mixers are available in multiple models and with a vast array of available platforms and accessories.
  • Available with a vast array of accessories
  • Digital Vortex-Genie features digital time and speed setting, touch or continuous mode
  • Two-year manufacturer's warranty
The Vortex Genie 1 touch mixer provides stronger handheld vortexing.
Optional accessories vastly increase the versatility of the vortexers. The TurboMix™ attachment provides hands-free, simultaneous, extremely vigorous vortexing and mixing of up to 12 microtubes. It isMore Product Information