AdjusTable® Workstation, Stainless Steel Solid Top, Stainless Steel Base, Eagle MHC™
Supplier: Eagle Group
AS2436T AS2448T AS2460T AS3036T AS3048T AS3060T
58202-840EA 3857.16 CAD
58202-840 58202-842 58202-844 58202-846 58202-848 58202-850
AdjusTable® Workstation, Stainless Steel Solid Top, Stainless Steel Base, Eagle MHC™
Furniture Tables and Workbenches
16 gauge, Type 304 brushed stainless steel solid top.
  • Stainless steel base with four 33" stainless posts
  • One stainless wire shelf and stainless C-frame
Load capacity: 600 lbs.
Order Now
SPECIFICATIONS
Environmentally Preferable