Utility Carts, Eagle MHC
Supplier: Eagle Group
EU2-1824C EU2-2136C EU2-2436Z U3-1836C U3-1842C U3-1842Z U2-2436C
58202-310EA 957.5 CAD
58202-310 58202-312 58202-314 58202-316 58202-318 58202-320 58202-322
Utility Carts, Eagle MHC
Trolleys
Shelves feature QuadTruss® design, making shelves up to 25% stronger and provides a retaining ledge for increased stability and product retention.
  • Optional cart configuration includes push or extended handle, four 33" (838 mm) posts, casters, and two or three shelves
  • Height includes casters
Medium and heavy-duty carts come with two or three wire shelves featuring patented QuadTruss® design, with EAGLEBrite® zinc, chrome, or stainless steel finish. Two handles, posts and casters with donut bumpers. Medium duty carts have 500 lbs. cart capacity. HeavyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable