VWR® 8" Test Sieves, Brass and Stainless Steel
Supplier: VWR International
57334-199 57334-201 57334-202 57334-208 57334-210 57334-212 57334-214 57334-216 57334-222 57334-224 57334-226 57334-228 57334-230 57334-232 57334-234 57334-236 57334-238 57334-240 57334-242 57334-244 57334-246 57334-248 57334-252 57334-254 57334-256 57334-258 57334-260 57334-262 57334-264 57334-268 57334-270 57334-272 57334-274 57334-276 57334-278 57334-280 57334-282 57334-284 57334-286 57334-288 57334-290 57334-292 57334-294 57334-296 57334-298 57334-300 57334-302 57334-304 57334-306 57334-308 57334-309 57334-310 57334-314 57334-324 57334-326 57334-328 57334-330 57334-334 57334-336 57334-339 57334-340 57334-342 57334-344 57334-346 57334-348 57334-350 57334-352 57334-354 57334-358 57334-360 57334-362 57334-364 57334-368 57334-370 57334-374 57334-376 57334-378 57334-380 57334-382 57334-384 57334-606 57334-610 57334-612 57334-614 57334-616 57334-618 57334-620 57334-622 57334-624 57334-626 57334-628 57334-630 57334-632 57334-634 57334-636 57334-200 57334-204 57334-206 57334-220 57334-250 57334-266 57334-356 57334-366 57334-372
57334-199EA 438.54 CAD
57334-199 57334-201 57334-202 57334-208 57334-210 57334-212 57334-214 57334-216 57334-222 57334-224 57334-226 57334-228 57334-230 57334-232 57334-234 57334-236 57334-238 57334-240 57334-242 57334-244 57334-246 57334-248 57334-252 57334-254 57334-256 57334-258 57334-260 57334-262 57334-264 57334-268 57334-270 57334-272 57334-274 57334-276 57334-278 57334-280 57334-282 57334-284 57334-286 57334-288 57334-290 57334-292 57334-294 57334-296 57334-298 57334-300 57334-302 57334-304 57334-306 57334-308 57334-309 57334-310 57334-314 57334-324 57334-326 57334-328 57334-330 57334-334 57334-336 57334-339 57334-340 57334-342 57334-344 57334-346 57334-348 57334-350 57334-352 57334-354 57334-358 57334-360 57334-362 57334-364 57334-368 57334-370 57334-374 57334-376 57334-378 57334-380 57334-382 57334-384 57334-606 57334-610 57334-612 57334-614 57334-616 57334-618 57334-620 57334-622 57334-624 57334-626 57334-628 57334-630 57334-632 57334-634 57334-636 57334-200 57334-204 57334-206 57334-220 57334-250 57334-266 57334-356 57334-366 57334-372
VWR® 8" Test Sieves, Brass and Stainless Steel
Sieves Test Sieves
VWR® 8" diameter (203.2 mm ) brass frame with stainless steel wire cloth test sieves are durable and ideal for sieving applications where a medium sample size is required.
  • Highly durable construction
  • Brass or stainless steel fabrication
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Order Now
SPECIFICATIONS
Environmentally Preferable