VWR® Test Sieves 8" , Brass
Supplier: VWR International
57334-100 57334-102 57334-106 57334-108 57334-112 57334-114 57334-116 57334-118 57334-120 57334-122 57334-124 57334-126 57334-130 57334-132 57334-134 57334-136 57334-138 57334-140 57334-142 57334-146 57334-150 57334-154 57334-156 57334-158 57334-160 57334-162 57334-164 57334-168 57334-172 57334-174 57334-176 57334-178 57334-180 57334-182 57334-184 57334-186 57334-188 57334-190 57334-192 57334-194 57334-196 57334-104 57334-110 57334-128 57334-144 57334-152 57334-166 57334-170
57334-100EA 350.82 CAD
57334-100 57334-102 57334-106 57334-108 57334-112 57334-114 57334-116 57334-118 57334-120 57334-122 57334-124 57334-126 57334-130 57334-132 57334-134 57334-136 57334-138 57334-140 57334-142 57334-146 57334-150 57334-154 57334-156 57334-158 57334-160 57334-162 57334-164 57334-168 57334-172 57334-174 57334-176 57334-178 57334-180 57334-182 57334-184 57334-186 57334-188 57334-190 57334-192 57334-194 57334-196 57334-104 57334-110 57334-128 57334-144 57334-152 57334-166 57334-170
VWR® Test Sieves 8" , Brass
Sieves Test Sieves
VWR® 8" diameter (203.2 mm ) all brass test sieves are durable and ideal for sieving applications where a medium sample size is required.
  • Highly durable construction
  • Brass fabrication
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are available as accessories.
Accessories information: Half height (1") or full heightMore Product Information