Sieves, Stainless Steel Wire, Brass Frame, W.S. Tyler™
Supplier: W.S. Tyler
6969 6989 7155 6990 6997 7163 6991 6998 7157 6992 7158 6993 7159 6994 7160 6995 7161 6996 7162 7164 6999 7165 7000 7166 7001 7167 7002 7003 7169 7004 7170 7005 7171 7006 7007 7173 7008 7174 7009 7010 7176 7011 7177 7012 7178 7013 7179 7014 7015 7181 7016 7182 7017 7183 6972 6974 6977 6979 6982 6984 6987
57332-080EA 503.2 CAD
57332-080 57332-084 57332-086 57332-088 57332-094 57332-096 57332-097 57332-098 57332-099 57332-104 57332-106 57332-108 57332-110 57332-112 57332-114 57332-116 57332-118 57332-120 57332-122 57332-124 57332-126 57332-128 57332-130 57332-132 57332-134 57332-136 57332-138 57332-142 57332-144 57332-146 57332-148 57332-150 57332-152 57332-154 57332-158 57332-160 57332-162 57332-164 57332-166 57332-170 57332-172 57332-174 57332-176 57332-178 57332-180 57332-182 57332-184 57332-186 57332-190 57332-192 57332-194 57332-196 57332-198 57332-200 57332-202 57332-204 57332-206 57332-208 57332-210 57332-212 57332-214
Sieves, Stainless Steel Wire, Brass Frame, W.S. Tyler™
Sieves Test Sieves
Full-height sieves are 8.3cm (31/4") high above stainless steel wire cloth
Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Sieve Number. Sieves one millimeter and coarser are identified by opening in millimeters and inches; sieves finer, by opening in microns and inches.
Overall height: 10.8cm (41/4"). Half-height sieves are 4.0cm (15/8") high above stainless steel wire cloth. Overall height: 6.6cm (25/8"). Diameter: 30.5cm (12").
Certifications: Conform to ASTM-E11 specifications.