Sampling Envelopes
Supplier: American Mail-Well Envelope
4056 4052 4060 4062
56775-029CS 1689.02 CAD
56775-029 56775-038 56775-041 56775-083
Sampling Envelopes
Bags Sample Bags
Made of extra-heavy tan kraft paper with improved metal fastener for storing mineral samples.