PIG® Conical Drain Plug, New Pig
Supplier: New Pig
PLR208 PLR209 PLR210 PLR252
56620-652EA 134.63 CAD
56620-652 56620-654 56620-656 56620-658
PIG® Conical Drain Plug, New Pig
Spill Control Materials
Durable polyurethane plug forms a liquid-tight, chemical-resistant seal in seconds.
Just wedge plug in drain for an easy alternative to permanently sealing your drains with cement. Installed plug is low-profile enough to fit below floor grate. Allows your drain to function normally when plug is not in place; removes easily with hand-tools.
Order Now
SPECIFICATIONS
Environmentally Preferable