Type I Safety Cans, Galvanized Steel, Eagle Manufacturing
Supplier: Eagle Manufacturing
UI4S UI10S UI10FS UI20S UI20FS UI50S UI50FS UI50SB UI50SY
56609-470EA 270.51 CAD
56609-470 56609-506 56609-517 56609-528 56609-550 56609-561 56609-572 56618-596 56618-598
Type I Safety Cans, Galvanized Steel, Eagle Manufacturing
Safety Cans
For carrying and dispensing flammable liquids safely.
Spout cap is controlled by a strong, nonrusting spring that holds the cap tight to prevent leakage or evaporation, but permits excessive gas pressure to escape and automatically closes cap when pressure is relieved. Pressing the open device allows liquid to be poured from the “no spill” pouring lip.
Made of heavy-gauge galvanized steel, finished in red baked-on powder coating. Brass flame arrester in spout absorbs and dissipates heat, keeping vapor temperature below flash point. Brass pour spout. Available in five standard Type IMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable