VWR® Biohazard Incinerator Cartons
Supplier: VWR International
56617-807 56617-810
56617-807PK 211.39 CAD
56617-807 56617-810
VWR® Biohazard Incinerator Cartons
Boxes Waste Disposal Boxes
A pair of tabs along the top edge of this cardboard receptacle keep the clear 0.038 mm (1.5 mil) thick polyethylene biohazard bag from slipping out of place.
  • Allows safe, convenient disposal and incineration of biohazardous waste
  • Negates the need to transfer contents
  • The snug-fitting lid has a flap for easily inserting waste material
Printed with large, bright, biohazard graphics and instructions in four languages (English, Spanish, French, German), it is packaged flat to save space and shipping costs. The carton and inner bag can be assembled inMore Product Information