Phenolic Foam Spill Control Pillows, NPS
Supplier: Avantor
56616-330CS 210.82 CAD
56616-330 56609-220
Phenolic Foam Spill Control Pillows, NPS
Spill Control Materials
Convenient pillows contain and absorb hazardous and toxic spills quickly
  • Able to absorb 98% of their capacity in 30 seconds
  • Pillows absorb a total of 10 to 20 times their weight of almost any liquid
  • Treat through elution, evaporation, or incineration in accordance with local regulations
Spill control pillows feature a nonpolluting, chemically inert glass foam to absorb acids, caustics, and organic solvents.
Caution: Not intended for use with strong oxidizers.