Nalgene® Benchtop Beta Waste Container (with bottle); Acrylic, PE bottle, Thermo Scientific
Supplier: Thermo Fisher Scientific
6710-2000
56616-035CS 847.39 CAD
56616-035
Nalgene® Benchtop Beta Waste Container (with bottle); Acrylic, PE bottle, Thermo Scientific
Bins Waste Bins
Nalgene Benchtop Beta Waste Container is constructed of 0.95 cm (3/8") thick acrylic
  • Constructed of 0.95 cm (3/8") thick acrylic
  • Cylindrical shield protects personnel from waste material
Provided with a 2-liter HDPE Nalgene wide mouth waste bottle for holding liquid wastes. Transparent shield encases the waste bottle without obstructing access to or manipulation of bottle closure. Machined lip on the cap prevents slipping and provides extra impact protection.
The cylindrical shield protects personnel from waste material contaminating beta-emitting isotopes.