I.C. Blue Lotion, R & R Lotion
Supplier: R & R Lotion
ICL-16-ESD ICL-8-ESD ICL-32 ICL-8
56614-106CS 481.69 CAD
56614-106 56614-118 56614-120 56616-116
I.C. Blue Lotion, R & R Lotion
Skincare Lotions Hand and Body Lotions
Blue I.C. pre-gloving, anti-static moisturizing lotion helps relieve dermatitis symptoms.
  • Greaseless and static dissipative
  • Contains no Silicones, Lanolins, Mineral Oil, or Glycerins
  • Non-contaminating
  • Can be used before gloving
I.C
Because dry skin can reduce the conductivity of grounding wrist straps, and this lotion moisturizes skin without introducing electrically resistive ingredients, it can help to meet American National Standards Institute/Electrostatic Discharge Association (ANSI/ESDA) S20.20 requirements. The lotion is available in aMore Product Information