Nonmetallic Dispenser Cans, Justrite®
Supplier: Justrite
14002 14004 14010
56609-740EA 125.76 CAD
56609-740 56609-744 56609-748
Nonmetallic Dispenser Cans, Justrite®
Safety Cans
For easy one-hand dispensing of flammable liquids directly onto work
Dispensers 56609-744 and -748 are FM approved.
Brass dispenser valves are leakproof and self-closing to minimize spills and control excess. Rugged polyethylene body.
Order Now
SPECIFICATIONS
Environmentally Preferable