VWR® High-Purity Two-Stage Gas Regulators, Stainless Steel
Supplier: VWR International
3300763 3300764 3300765 3300766 3300767 3300768 3300769 3300770 3300771 3300772 3300773 3300774
55850-674EA 3895.06 CAD
55850-674 55850-676 55850-678 55850-680 55850-682 55850-684 55850-686 55850-688 55850-690 55850-692 55850-694 55850-696
VWR® High-Purity Two-Stage Gas Regulators, Stainless Steel
Gas Regulators
Regulators have a stainless steel barstock body and are suitable for corrosive gas applications, high-purity gas applications, research sample systems, process analyzers, gas chromatography, EPA protocol mixes, laser gas systems, and emission monitoring systems.
  • Suitable for high-purity applications
  • Two-stage design for constant outlet pressure
  • Stainless steel diaphragm prevents contaminations
  • One-piece PTFE encapsulated seat design with sintered filter
The two-stage design provides constant outlet pressure regardless of change in cylinder pressure.More Product Information