VWR® High-Purity Single-Stage Gas Regulators, Stainless Steel
Supplier: VWR International
3001156 3001157 3001166 3001167 3001158 3001159 3001160 3001161 3001162 3001163 3001164
55850-650EA 2961.92 CAD
55850-650 55850-652 55850-654 55850-656 55850-658 55850-660 55850-662 55850-664 55850-666 55850-668 55850-670
VWR® High-Purity Single-Stage Gas Regulators, Stainless Steel
Gas Regulators
Regulators have a stainless steel barstock body and are suitable for a variety of applications where a rise in delivery pressure from a full to an empty cylinder can be tolerated.
  • Stainless steel body
  • Stainless steel diaphragm
  • Sintered filter
  • PTFE encapsulated seat design
They may be used with corrosive gas applications
Other delivery pressures and outlet connections are available; contact your Avantor sales representative for information.
They are also ideal for high-purity gas applications, research sample systems, process analyzers, gasMore Product Information