VWR® Multistage Gas Regulators with Neoprene Diaphragms
Supplier: VWR International
55850-388 55850-390 55850-392 55850-396 55850-398 55850-472 55850-474 55850-476 55850-478 55850-480 55850-482 55850-484 55850-486 55850-488 55850-490 55850-492 55850-494 55850-496 55850-498
55850-388EA 1033.34 CAD
55850-388 55850-390 55850-392 55850-396 55850-398 55850-472 55850-474 55850-476 55850-478 55850-480 55850-482 55850-484 55850-486 55850-488 55850-490 55850-492 55850-494 55850-496 55850-498
VWR® Multistage Gas Regulators with Neoprene Diaphragms
Gas Regulators
VWR® Multistage Gas Regulators with Neoprene Diaphragms are designed for applications where a constant working pressure is important over a wide range of inlet pressures and flow rates.
  • Solderless construction for easy maintenance
  • Solid forged brass body contains two regulators
  • First stage reduces full cylinder pressure to 90%
  • Sintered inlet filter removes particulates, tank scale, and rust from gas stream
VWR® Multistage Gas Regulators feature safe, accurate, and sensitive delivery of reduced pressure
Note: For noncorrosive gases only.