VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Supplier: VWR International
55850-416 55850-418 55850-420 55850-422 55850-424 55850-426 55850-428 55850-432 55850-434 55850-438 55850-440 55850-442 55850-444 55850-446 55850-450 55850-452 55850-456 55850-458 55850-460 55850-462 55850-464 55850-468 55850-412 55850-430 55850-436 55850-414
55850-416EA 1199.39 CAD
55850-416 55850-418 55850-420 55850-422 55850-424 55850-426 55850-428 55850-432 55850-434 55850-438 55850-440 55850-442 55850-444 55850-446 55850-450 55850-452 55850-456 55850-458 55850-460 55850-462 55850-464 55850-468 55850-412 55850-430 55850-436 55850-414
VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Gas Regulators
VWR® Multistage gas regulators with stainless steel diaphragms are designed for gas chromatography applications where a constant working pressure is important over a wide range of inlet pressures and flow rates.
  • Solderless construction for easy maintenance
  • Solid forged brass body contains two regulators
  • First stage reduces full cylinder pressure to 90%
  • Sintered inlet filter removes particulates, tank scale, and rust from gas stream
  • Stainless steel diaphragms
  • Supply pressure gauge: 4000 psig
VWR® Multistage Gas Regulators feature safe, accurate, andMore Product Information