Nalgene® CryoBox™ Boxes, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
5025-0505 5026-0909 5027-0909
55709-946CS 873.34 CAD
55709-946 55709-968 55709-972
Nalgene® CryoBox™ Boxes, Polycarbonate, Thermo Scientific
Boxes Cryoboxes
For ultra-low temperature storage of both internally and externally threaded cryogenic vials
Durable, economical alternative to flimsy cardboard or expensive stainless steel storage boxes. Polycarbonate has usable temperature of –196 to 121°C. Numerical grid system features molded-in numbers on box and corresponding numbers printed in blue on lid. Clear lids are keyed in one position so grids always match. Boxes accept writing with markers designed for ultra-low temperatures. Fits the Thermolyne® Locator® Cryobiological Storage Vessels.
Ideal for use in mechanical and liquid-nitrogenMore Product Information