VWR® CryoPro® V Vapor Shippers
Supplier: VWR International
55709-226 55709-230 55709-232 55709-228
55709-226EA 4930.48 CAD
55709-226 55709-230 55709-232 55709-228
VWR® CryoPro® V Vapor Shippers
Cryopreservation Systems
Using the latest technology in hydrophobic absorbents, CryoPro® vapor shippers are designed for safe shipment of biological specimens at cryogenic temperatures.
  • Moisture-repelling properties
  • Optional shipping cases available
  • Sealed shaft prevents hazardous spurting of liquid nitrogen
Moisture-repelling absorbent allows for maximum hold times, shipment after shipment.
CryoRetriever claw-style tool (82017-944) is specifically designed for retrieving boxes, vials, and frames from cryopreservation tanks. Exclusive sealed center shaft prevents hazardousMore Product Information