Refractometer, Industrial Fluid Tester, Reichert
Supplier: Reichert Technologies
13754000
55680-389EA 727.65 CAD
55680-389
Refractometer, Industrial Fluid Tester, Reichert
Refractometers Handheld Refractometers
Automatic temperature compensation for immediate, accurate analysis without referencing temperature conversion charts
Easy-to-read scale instantly measures cutting fluids, water soluble coolants, oil/water mixtures, cleaners, plating baths, and many other types of industrial solutions. Includes instructions for maintaining optimum fluid/water ratios. Durable, water-resistant design. Scale is measurement of an arbitrary scale based on coolants for metal working.