DRYFAST®, DRYFAST ULTRA®, and DRYFAST COLLEGIATE Chemical Duty Vacuum Pumps with Advanced Vapor Management, Welch®
Supplier: Welch Vacuum
2032B-01 2042B-01 2044B-01 2037B-01 2032C-02 2044C-02 2014B-01 2014C-02 2034B-01 2047B-01
55009-284EA 9132.04 CAD
55009-284 55009-286 55009-290 55009-292 55009-296 55009-298 55009-302 55009-304 55009-306 97025-730 97025-732 55009-288 55009-294
DRYFAST®, DRYFAST ULTRA®, and DRYFAST COLLEGIATE Chemical Duty Vacuum Pumps with Advanced Vapor Management, Welch®
Pumps Vacuum Pumps
These dry vacuum pumps, designed to meet chemical duty laboratory needs, are ideal for rotary evaporation, concentrators, evaporators, vacuum ovens, filtration, degassing, and other chemical applications
Pumps are constructed with solid PTFE heads, PTFE diaphragms, Kalrez® valves, and utilize fluorinated plastics on all wetted surfaces. Pumps feature flows to 70L/min and ultimate vacuum levels down to 2torr. No cold traps are needed to operate. These stand-alone vacuum pumps need very little maintenance, leading to low cost of ownership and improved productivity.
Pumps feature built-inMore Product Information