Reducers, Welch®, Gardner Denver
Supplier: Welch Vacuum
387121 387132
55009-238EA 242.62 CAD
55009-238 55009-191
Reducers, Welch®, Gardner Denver
Fittings and Connectors
Used to mate two different sizes of ISO flanges
Made of stainless steel.
Order Now
SPECIFICATIONS
Environmentally Preferable