DUOSEAL® Belt-Drive Vacuum Pumps, Two Stage, Welch®
Supplier: Welch Vacuum
1400C-02 1405C-02 1402C-02 1400B-01 1400 1405B-01 1402B-01 1402W-01 1402 1376B-01 1374B-01 1400B-80 1400C-01 1400W-01 1405B-80 1405W-01 1402B-46 1402B-80 1397B-01 1397M-01 1374M-01
54970-602EA 4900.54 CAD
54970-602 54970-604 54970-606 54973-075 54973-133 54974-238 54976-029 54976-053 54976-110 54987-070 54993-505 54973-084 54973-087 54973-213 54974-247 54974-262 54976-035 54976-038 54993-345 54993-360 54993-517
DUOSEAL® Belt-Drive Vacuum Pumps, Two Stage, Welch®
Pumps Vacuum Pumps
Use these rugged oil-seal pumps for a wide range of laboratory vacuum needs
Durable by design, DUOSEAL® pumps are built with precisely machined components to rigorous quality standards. Low pump rpm operation reduces friction, oil temperature, and oil degradation to extend maintenance intervals and pump life. Large oil reservoir minimizes contaminant effects to maintain high vacuum performance. Use of a cold trap is recommended to protect the pump and enhance vacuum levels. Do not operate continuously at pressures greater than 10 torr.
DUOSEAL® vacuum pump applications include SchlenkMore Product Information