SP Bel-Art Vari-Pet® Pipettors, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37895-0010 F37895-1100
54848-208EA 693.59 CAD
54848-208 54848-218 54848-222 54848-212
SP Bel-Art Vari-Pet® Pipettors, Bel-Art Products, a part of SP
Pipettes Micropipettes
This self-filling pipetting aid is designed for speed, ease, and convenience.
  • Polyproyplene handle
  • Teflon®-coated borosilicate glass pipettor
  • Ergonomically designed for extended use
  • Adjustable dispensing volume
The polypropylene plastic handle is ergonomically designed to permit extended use without fatigue and a Teflon® plunger assures smooth, consistent operation.
Teflon® Tip Vari-Pet® Pipettor has excellent chemical resistance because the fluids only contact borosilicate glass and Teflon®.
Luer Lock Vari-Pet® Pipettor is supplied with aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable