HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Supplier: Air-Tite
4010.200V0 4020.000V0 4050.000VZ 4100.000V0 4200.000V0 NJ-9166017-02 NJ-4606027-02 NJ-4606067-02 NJ-4606108-02 NJ-4606205-02
53548-001CS 844.42 CAD
53548-001 53548-003 53548-005 53548-024 53548-006 53548-008 53548-010 76459-474 76459-476 76459-478 76459-480 76459-482
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Syringes Syringes, Medical
NORM-JECT® syringes feature medical-grade plastics, including a polypropylene barrel and a polyethylene plunger. Great value at reasonable costs.
  • Sterile
  • Certified DNA free
  • Latex free
  • Rubber free
  • Silicone free
  • Styrene free
  • DEHP free
These syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition, these syringes are indicated for Nuclear Medicine, Amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. TheseMore Product Information