MLA Precision Pipettors, VistaLab
Supplier: VistaLab
1021 1027 1108 1104 1115 1025 1028 1029 1101 1107 1106 1024 1112 1109 1117 1118 1022 1023 1026 1113 1119 1103 1030 1110 1102 1111 1505
53514-004EA 568.56 CAD
53514-004 53514-016 53514-066 49000-518 49000-524 53514-012 53514-018 53514-020 53514-062 53514-064 49000-522 53514-010 53514-070 49000-526 49000-530 49000-532 53514-006 53514-008 53514-014 53514-072 53514-074 49000-514 53514-022 53514-068 75813-802 75813-804 75814-378
MLA Precision Pipettors, VistaLab
Pipettes Repetitive Pipettors
MLA pipettors are made of precision-engineered aluminum alloy and stainless steel, and are designed for highly accurate delivery and proven performance.
  • Available in a wide range of fixed volumes
  • Light plunger forces for easy operation with thumb/forefinger
  • Single-stroke 'to deliver' operation
  • Individually serialized
  • Lifetime warranty on parts
These pipettors are available in 28 fixed volumes ranging from 5 to 1000 μL. Pipettors can be easily adjusted as needed to achieve accurate performance on dense or viscous liquids.
Accessories information: More Product Information