Upchurch Scientific® Stainless Steel Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
U-153 U-157 U-1120 U-104 U-105 U-111 U-114 U-118 U-131 U-150 1321 U-135 U-136 U-143 U-144 1323 U-164 U-102 U-132 U-152 U-154 1322 1324 U-103 U-106 U-107 U-113 U-116 U-117 U-126 U-127 U-128 U-133 U-140 U-162 U-163 U-800 1320 U-115 U-134 U-137 U-155 U-156 U-160 U-165 U-803 U-101 U-108 U-112 U-129 U-139 U-141 U-158 U-1114
21511-286EA 17.44 CAD
21511-286 53500-718 97055-120 21511-240 21511-242 21511-250 21511-256 21511-262 21511-272 21511-284 21511-356 53500-696 53500-698 53500-704 53500-706 53500-708 53500-724 21511-236 21511-274 53500-710 53500-712 21001-100 21001-102 21511-238 21511-244 21511-246 21511-254 21511-258 21511-260 21511-264 21511-266 21511-268 21511-276 21511-280 21511-288 21511-290 21511-344 21511-354 53500-692 53500-694 53500-700 53500-714 53500-716 53500-722 53500-726 53500-758 21511-234 21511-248 21511-252 21511-270 21511-278 21511-282 53500-720 97055-118
Upchurch Scientific® Stainless Steel Tubing, IDEX Health & Science
Tubing Transfer Tubing
Upchurch Scientific® seamless, precut, stainless steel tubing is designed to meet the exacting requirements of today's analyses.
  • Autoclavable
  • Sterilization: Gamma irradiation; ethylene oxide; thermal
A variety of precut lengths as well as longer lengths (1 to 7.6 m) of some sizes are available. Cutting the tubing disturbs and roughens the tubing's end surface, so it is recommended to use precut tubing lengths whenever possible. If custom lengths are required, it is suggested that the tubing is passivated after cutting. Stainless steel precut tubing isMore Product Information