Microcaps® Disposable Micropipettes, Drummond
Supplier: Drummond
1-000-2000-E 1-000-00025 1-000-0005 1-000-0010-64 1000002055 1-000-0030 1000003055 1-000-0040 1-000-0060 1-000-0080 1-000-0150 1-000-0090 1-000-0250 1-000-0300 1-000-0400 1-000-0447 1-000-0750 1-000-0002 1000001055 1-000-0033 1000005055 1-000-0066 1-000-0070 1000020032 1-000-0350 1000040080 1-000-2000 1-000-9000 1-000-0005-64 1-000-0050/CA
47729-742EA 89.08 CAD
47729-742 53439-964 53439-986 53440-005 53440-023 53440-027 53440-045 53440-049 53440-067 53440-090 53440-114 53440-125 53440-136 53440-147 53440-169 53440-180 53440-205 53440-216 53440-227 53440-249 53440-260 53507-102 53507-104 53507-122 53507-132 53507-138 53507-146 53507-148 53507-172 53507-192 53507-196 53507-236 53507-268 53440-001 53440-089 53440-103 490007-158 53440-088
Microcaps® Disposable Micropipettes, Drummond
Pipettes Microcapillaries
Drummond microcaps are a fast, safe, clean, and accurate way to transfer liquids in micro quantities.
  • Accurate (±1%)
  • Dependable
  • Easy to use
  • Fast
They are precision-bore glass capillary tubes cut to predetermined lengths, so fill volumes are constant and precise.
Capillary action draws the fluid into the tube, and a squeeze of the bulb dispenses it. For spotting applications such as TLC, a provided spotting holder allows the liquid to be dispensed by touching the tube to the spotting surface.
Microcaps are inexpensive and disposable.