Disposable Standard-Tip Serological Pipettes, Plastic, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
56800-21100 56800-10110 56825-10110 56700-11100 56700-10110 56800-25210 56700-21100
14672-902CS 1219.18 CAD
14672-902 14672-906 14672-908 14672-910 14672-912 53384-325 53384-331 53384-443 53384-447 72700-100
Disposable Standard-Tip Serological Pipettes, Plastic, Kimble Chase
Pipettes Serological Pipettes
Standard-Tip Serological Pipettes are irradiation sterilized and suitable for tissue culture work.
They are equipped with uniform pulled or molded tips; beveled or molded top ends with cotton plugging. Two rings near the top denote that the small amount remaining in tip after delivery must be blown out and added to main volume delivered. The 5 ml size features a two-piece design. Color-coded packaging makes choosing the correct pipette size quick and accurate. Each pipette is equipped with negative and reverse graduations.
Certifications: Designed from ASTM E 934. Calibrated 'ToMore Product Information