ART® XLP 200 Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Supplier: Thermo Scientific
2160P
53225-800CS 1909.21 CAD
53225-800
ART® XLP 200 Self-Sealing Barrier Pipette Tips, Molecular BioProducts
Pipette Tips
Tips ensure 100% protection against aerosol and liquid contamination.
  • Pre-sterilized tips
These tips feature an extended tip length like the ART® XLP but also draw a full 200 µl of volume. The ART® barrier traps contaminants, keeping them out of pipettors and samples. This also protects the sensitive inner components of the pipettor, prolonging pipettor life. ART® tips are ideal for nucleic amplification procedures, pipetting of radioactive samples, or any application where critical sample handling is required. Tips are compatible with Eppendorf®,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO14001 (Certificate Registration No. EMS 752603)