PYREX® Reusable Volumetric Pipettes, Class A, Certified/Serialized, Corning
Supplier: Corning
7070-1 7070-2 7070-5 7070-10 7070-25
53044-730CS 373.45 CAD
53044-730 53044-732 53044-734 53044-736 53044-738
PYREX® Reusable Volumetric Pipettes, Class A, Certified/Serialized, Corning
Pipettes Measuring (Mohr) Pipettes
These reusable PYREX® glass pipettes are colour-coded by size for easy identification and sorting.
  • Calibrated 'to deliver' (TD) their total capacity without blow-out
  • Top end of 5 to 25 ml sizes is constricted (Mohr type)
  • Coloured graduations are enameled onto the glass
  • Colour-coded by size for easy identification
  • Each pipette is individually serialised and certified
PYREX® Serialized/Certified Volumetric Glassware is now tested and calibrated in an ISO/IEC 17025 certified laboratory.
Certifications: Calibrated to Class A tolerances in accordanceMore Product Information