CaptairBio PCR Workstations, Erlab
Supplier: Erlab
321PCR 391PCR 321SST 391SST
52428-824EA 7253.78 CAD
52428-824 52428-826 10170-026 10170-030
CaptairBio PCR Workstations, Erlab
Fume Hoods and Enclosures PCR Workstation Enclosures
Erlab’s CaptairBio PCR Workstations are designed to protect sensitive biological applications against both environmental pollution and cross-contamination. The HEPA filter will bring clean air down to the work surface, with high efficiency UV lights to decontaminate the worktop from biological cross-contamination.
  • Each unit has Smart Technology™ - an exclusive set of tools that consists of Smart-Light Communication™, chemical sensors (*optional if adding carbon filters), real-time status, and the eGuard app
  • Smart Technology gives an easily identifiable
More Product Information