VWR® Critical Print™ Scientific Notebooks
Supplier: VWR International
51128-000 51280-856
51128-000CS 1268.96 CAD
51128-000 51280-856
VWR® Critical Print™ Scientific Notebooks
Notebooks
Hardbound notebooks feature a title page and a table of contents.
  • Low-particulating paper
  • Numbered pages
  • College ruled or grid style
Each page has space for signature, witness, and date. Notebooks include 200 numbered pages (100 sheets) of low particulating paper. Choose from college ruled or 6.4mm (1/4") grid style.
Ordering information: A4-sized notebooks are also available, in both college ruled and grid styles; contact your Avantor sales representative for more information.