Cole-Parmer® EH-200 Series Macro-Kjeldahl Extraction Heaters, Antylia Scientific
Supplier: Antyila Scientific
03075-00
51035-020EA 6513.31 CAD
51035-020
Cole-Parmer® EH-200 Series Macro-Kjeldahl Extraction Heaters, Antylia Scientific
Heaters Heating Mantles
Designed to determine the protein content of food on a macro scale - from 100 to 800 ml.
  • Independent heat controller lets you adjust the temperature range of each mantle individually
  • Stainless steel construction is rugged and easy to clean
Run several projects simultaneously using these multiposition macro-kjeldahl extraction heaters. Heaters are available with two or six individually controlled heating mantles that accommodate either 100 to 300 ml vessels or 500 to 800 ml vessels. Back-mounted brackets hold flask support rods up to 12.7 mm inMore Product Information