PEEK Sample Loops, Rheodyne®
Supplier: Rheodyne
9055-021 9055-022 9055-023 9055-024 9055-025 9055-026 9055-027 9055-028 9055-029 9055-020
50807-100EA 190.03 CAD
50807-100 50807-102 50807-104 50807-106 50807-108 50807-110 50807-112 50807-114 50807-116 50807-516
PEEK Sample Loops, Rheodyne®
Chromatography Supplies Chromatography Sample Loops
These loops are designed for Rheodyne models 9125, 9725, and 9010 PEEK injectors.
Loops are supplied with fitting that are unswaged so that the tube may be completely bottomed in the injector port before the ferrule is swaged on. Accuracy of large loops (0.030" bore) is about 14%, intermediate loops (0.020") 21%, and small loops (0.007") 65%. The size designations are nominal, and the actual volumes can differ from the nominal designations due to the ±0.002" tolerance of the tubing bore.
Order Now
SPECIFICATIONS
Environmentally Preferable