High-Pressure Syringes, Antron Engineering
Supplier: Antron Engineering
PRO-7300 PRO-7300-14 PRO-7300-18 PRO-7300-16 PRO-7300-LL PRO-7400 PRO-7400-14 PRO-7400-18 PRO-7400-16 PRO-7400-LL PRO-7500 PRO-7500-14 PRO-7500-18 PRO-7500-16 PRO-7500-LL PRO-7600 PRO-7600-14 PRO-7600-18 PRO-7600-16 PRO-7600-LL PRO-7100 PRO-7100-18 PRO-7100-16 PRO-7100-LL PRO-7700 PRO-7700-18 PRO-7700-16 PRO-7700-LL
50000-454EA 2093.64 CAD
50000-454 50000-456 50000-458 50000-460 50000-462 50000-464 50000-466 50000-468 50000-470 50000-472 50000-474 50000-476 50000-478 50000-480 50000-482 50000-484 50000-486 50000-488 50000-490 50000-492 50000-494 50000-496 50000-498 50000-500 50000-502 50000-504 50000-506 50000-508
High-Pressure Syringes, Antron Engineering
Syringes General-Purpose Syringes
These syringes are intended for high-pressure applications and offer good resistance to most aggressive liquids
Accessory replacement O-ring kits include two piston rings and one tip ring. They are replacements to be used with Antron Engineering syringes (see 50000-494 series). Kalrez® perfluoroelastomer O-rings feature exceptional resistance to degradation by aggressive fluids and gases. Viton® fluoroelastomer O-rings offer excellent resistance to aggressive fuels and chemicals.
Syringe barrels and plungers are made of type 316 stainless steel. Syringes are leak-free to pressures upMore Product Information