J.T.Baker® Robotic Tips Type TE MCA
Supplier: Ritter, part of Avantor
49302-00081 49302-00091 49302-10081 49302-11091 49304-00081 49304-00091 49304-10081 49304-11091 49305-00091 49306-00081 49306-00091 49306-10081 49306-11091 49332-00091
49302-0008CS 672.01 CAD
49302-0008 49302-0009 49302-1008 49302-1109 49304-0008 49304-0009 49304-1008 49304-1109 49305-0009 49306-0008 49306-0009 49306-1008 49306-1109 49332-0009
J.T.Baker® Robotic Tips Type TE MCA
Pipette Tips
J.T.Baker Robotic tips type TE MCA are compatible with the Tecan Freedom EVO and the Tecan Fluent liquid handler using the MCA 96 head. Tips are manufactured under automated processes to reduce human contact and ensure precise reproducibility and exceptional quality.
  • Tested and certified free of human DNA, DNase and RNase, BSE/TSE, PCR inhibitors, ATP, and heavy metals
  • Manufactured in an ISO 9001 & ISO 13485 registered facility under cleanroom (class 8) conditions
  • Tips are made with virgin polypropylene ensuring your lab automation is free from resin
More Product Information