J.T.Baker® Robotic Tips Type TE
Supplier: Ritter, part of Avantor
49002-00061 49002-01061 49002-11061 49003-00061 49003-01061 49003-11061 49006-00061 49006-11061 49033-00061 49002-00001 49003-00001 49006-00001 49033-00001 49102-00001 49102-00061 49102-11061 49103-00001 49006-01061 49033-01061 49103-00061 49103-11061 49106-00001 49106-11061 49133-00001 49024-00081 49033-11061 49026-00171 49026-01171 49026-11171
49002-0006CS 557.66 CAD
49002-0006 49002-0106 49002-1106 49003-0006 49003-0106 49003-1106 49006-0006 49006-1106 49033-0006 49002-0000 49003-0000 49006-0000 49006-0107 49033-0000 49102-0000 49102-0006 49102-1106 49103-0000 49006-0106 49033-0106 49103-0006 49103-1106 49106-0000 49106-1106 49133-0000 49024-0008 49006-1107 49033-1106 49026-0017 49026-0117 49026-1117
J.T.Baker® Robotic Tips Type TE
Pipette Tips
Suitable for use on Tecan Freedom EVO or Tecan Fluent. Available in conductive and non-conductive version, with or without filter.
  • Manufactured under cleanroom conditions (cleanroom class 8)
  • State of the art quality control
  • Ideal for high-throughput analysis
  • Packed in cardboard or blister
J.T.Baker® robotic tips type TE are suitable for use on Tecan Freedom EVO (typically uses tips in S-Tray = hanging tray), Tecan Fluent (typically uses tips in C-Tray for ANSI/SLAS format tip box) and other OEM robotic instruments.
The validation of the tips on theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)