D-(+)-Trehalose dihydrate 99,0, reagent

Supplier: VWR

Synonyms: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

K480-25G K480-100G
490015-644EA 193.57 CAD
490015-644 10118-236
D-(+)-Trehalose dihydrate 99,0, reagent
D(+)-Trehalose dihydrate
Formula: C₁₂H₂₂O₁₁·2H₂O
MW: 378.33 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00071594
CAS Number: 6138-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chloride 0.018%
Heavy Metals (as Pb) = 3 ppm
Nitrogen = 0.01%
pH (30%, Water) @25°C 4.5 - 6.5
Purity = 99%
Residue on Ignition = 0.05%
Sulfate = 0.024%
Water (KF) = 11.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR