Nylon Cloth Wire and Lab Labels, 3" Core, Brady
Supplier: Brady Worldwide
30542 THT-165-499-5
490006-386EA 700.78 CAD
490006-386 76461-156
Nylon Cloth Wire and Lab Labels, 3" Core, Brady
Labels
Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
  • Autoclavable material
  • Suitable for cryogenic storage
  • Service temperature of ‒40 to +90° C (‒40 to +193 °F)
Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable. Recommended for indoor use.
Labels are designed for use with BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, orMore Product Information