Kimble® KIMAX® Cream Pipettes with Wide Tip, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
570-9
48756-028CS 387.26 CAD
48756-028
Kimble® KIMAX® Cream Pipettes with Wide Tip, DWK Life Sciences
Pipettes Milk Testing Pipettes
Reusable glass pipettes with wide tip.
  • Scale is permanent brown stain
  • Calibrated to deliver
Designed from ASTM Specification E1043, Type III