KIMAX® Babcock Bottle, Skim Milk Test, 0.5%, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
530-50100
48717-004CS 1304.98 CAD
48717-004
KIMAX® Babcock Bottle, Skim Milk Test, 0.5%, Kimble Chase
Bottles Dairy Testing Bottles
KIMAX® bottle with auxiliary filling tube on side to fill bottle.
Graduated neck has too narrow a bore to fill through. Neck is graduated to 0.5% in 0.01% subdivisions. Scale is permanent brown stain.