SP Bel-Art CryoSafe Mini Cooler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18846-0000
47751-726EA 353.18 CAD
47751-726
SP Bel-Art CryoSafe Mini Cooler, Bel-Art Products, a part of SP
Coolers Benchtop Coolers
This benchtop sized cooler holds twelve 0.5 or 1.5 ml microcentrifuge tubes at a temperature below 0 ºC for approximately 2.5 hours.
  • Polycarbonate cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe
  • Clear polycarbonate lid shows positions of tubes; keyed to correspond with positions printed on cooler
  • Molded side handles allow secure grip for transfer
  • Rubber feet to prevent sliding on the lab bench
  • Stackable
Ideal for protecting valuable samples stored in a freezer with defrost cycle.