Axygen® 96-Well PCR Plates Scientific
Supplier: Corning
PCR-96-C PCR-96-C-S PCR-96-FS-C PCR-96-HS-C PCR-96-AB-C
10011-220CS 991.81 CAD
10011-220 10011-222 10011-228 10011-248 10011-266 47749-876
Axygen® 96-Well PCR Plates Scientific
Microplates PCR Plates
PP plates, silicone mats. Designed to fit 0.2 ml thermal cycler blocks.
  • Ultra-thin walls of uniform thickness accelerate heat transfer, leading to shorter cycle times and more consistent results
  • Certified free from DNase, RNase and endotoxin for highest sample integrity
  • AxyMat™ sealing mats autoclavable and reusable
Volume with AxyMat™: 240 μl
Compatible with (in addition to standard thermal cycling): ABI Fast PCR 5700, 7000, 7700, BioRad iCycler, MyiQ, iQ5, ABI sequencers.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Reynosa