Qualitative Filter Paper, Grade 610, Ahlstrom
Supplier: Ahlstrom
6100-0900 6100-1100 6100-1850 6100-0750 6100-2400 6100-0250 6100-0425 6100-0700 6100-2500 6100-2700 6100-3200 6100-5000 6100-0550 6100-1250 6100-1500
38005-068BX 34.06 CAD
38005-068 38005-080 38005-148 47745-800 47745-802 47749-672 47749-674 47749-676 47749-678 47749-680 47749-682 47749-684 38005-024 38005-104 38005-126
Qualitative Filter Paper, Grade 610, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 610 is a slow flowing filter designed to retain fine particles.
It is widely used for water analysis, biological products and clarifying cloudy suspensions. This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.
Order Now
SPECIFICATIONS
Environmentally Preferable
Flow rate Slow
Load capacity Medium
Retention 1.5 µm