Qualitative Filter Paper, Grade 601, Ahlstrom
Supplier: Ahlstrom
6010-0700 6010-0425 6010-1850 6010-2400 6010-3200 6010-0550 6010-0750 6010-0900 6010-1100 6010-1250 6010-1500 6010-0250 6010-0300 6010-0470 6010-2500 6010-2700 6010-3850 6010-4000 6010-5000
28342-016BX 18.04 CAD
28342-016 38000-025 38000-160 38000-182 38000-229 47745-808 47745-810 47745-812 47745-814 47745-816 47745-818 47749-656 47749-658 47749-660 47749-662 47749-664 47749-666 47749-668 47749-670
Qualitative Filter Paper, Grade 601, Ahlstrom
Filters Filter Papers Qualitative Filter Paper
Ahlstrom Grade 601 is used in a wide range of laboratory applications such as clarifying liquids, rapid filtration of coarser precipitates, soil analysis, seed testing and more.
This low ash grade contains more than 95% super refined alpha-cellulose and linter fibers.
Order Now
SPECIFICATIONS
Environmentally Preferable
Flow rate Medium
Load capacity Medium
Retention 2.5 µm