Ashless Quantitative Paper, Grade 94, Ahlstrom
Supplier: Ahlstrom
0940-0550 0940-1100 0940-0900 0940-1250 0940-1500 0940-0425 0940-0700 0940-1850 0940-2400 0940-4000 0940-0470
38009-048BX 59.76 CAD
38009-048 38009-106 47745-754 47745-756 47745-758 47749-498 47749-500 47749-502 47749-504 47749-506 30621-280
Ashless Quantitative Paper, Grade 94, Ahlstrom
Filters Filter Papers Quantitative Filter Paper
Ahlstrom Grade 94 is an extremely fine retention ashless (Ash 0.007%) filter paper suitable for routine quantitative gravimetric techniques.
Order Now
SPECIFICATIONS
Environmentally Preferable
Flow rate Slow
Load capacity Medium
Retention 1.5 µm